Assay Details
| Gene Symbol: Oxct1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024188.6 | NP_077150.1 | 6 - 7 | 1013 | 59 | |
| GenBank mRNA | AB085609.1 | - | 6 - 7 | 755 | 59 | |
| AK009027.1 | - | 6 - 7 | 793 | 59 | ||
| AK010790.1 | - | 6 - 7 | 756 | 59 | ||
| AK011354.1 | - | 6 - 7 | 693 | 59 | ||
| AK078086.1 | - | 6 - 7 | 1013 | 59 | ||
| AK146158.1 | - | 6 - 7 | 771 | 59 | ||
| AK146342.1 | - | 6 - 7 | 758 | 59 | ||
| AK151662.1 | - | 6 - 7 | 752 | 59 | ||
| AK151695.1 | - | 6 - 7 | 743 | 59 | ||
| AK167681.1 | - | 6 - 7 | 843 | 59 | ||
| BC003422.1 | - | 6 - 7 | 717 | 2598824 | 59 | |
| EF095212.1 | - | 3 - 4 | 406 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Oxct1
Genomic Map