Assay Details
| Gene Symbol: Xrcc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010247.2 | NP_034377.2 | 1 - 2 | 102 | 109 | |
| GenBank mRNA | Z14157.1 | - | 240 | 109 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_010247.2, Z14157.1 transcripts.
Set Membership: |
|

Xrcc6
Genomic Map