Assay Details
| Gene Symbol: Xrcc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010247.2 | NP_034377.2 | 3 - 4 | 304 | 96 | |
| GenBank mRNA | AB010282.1 | - | 3 - 4 | 245 | 96 | |
| AF483500.1 | - | 2 - 3 | 184 | 96 | ||
| AF483501.1 | - | 2 - 3 | 184 | 96 | ||
| AK143570.1 | - | 3 - 4 | 279 | 96 | ||
| AK146392.1 | - | 3 - 4 | 223 | 96 | ||
| AK146394.1 | - | 3 - 4 | 223 | 96 | ||
| AK146396.1 | - | 3 - 4 | 271 | 96 | ||
| BC031422.1 | - | 3 - 4 | 253 | 3669715 | 96 | |
| BC051085.1 | - | 3 - 4 | 1482 | 96 | ||
| BY292222.1 | - | 3 - 4 | 306 | 96 | ||
| M38700.1 | - | 2 - 3 | 146 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Xrcc6
Genomic Map