Assay Details
| Gene Symbol: Xrcc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010247.2 | NP_034377.2 | 9 - 10 | 1402 | 69 | |
| GenBank mRNA | AB010282.1 | - | 9 - 10 | 1343 | 69 | |
| AF483500.1 | - | 8 - 9 | 1282 | 69 | ||
| AF483501.1 | - | 8 - 9 | 1282 | 69 | ||
| AK143570.1 | - | 9 - 10 | 1377 | 69 | ||
| AK146392.1 | - | 9 - 10 | 1321 | 69 | ||
| AK146394.1 | - | 9 - 10 | 1321 | 69 | ||
| AK146396.1 | - | 9 - 10 | 1369 | 69 | ||
| BC031422.1 | - | 9 - 10 | 1351 | 3669715 | 69 | |
| BC051085.1 | - | 9 - 10 | 2580 | 69 | ||
| M38700.1 | - | 8 - 9 | 1244 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Xrcc6
Genomic Map