Assay Details
| Gene Symbol: Slc38a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166456.1 | NP_001159928.1 | 13 - 14 | 1428 | 79 | |
| NM_001166458.1 | NP_001159930.1 | 13 - 14 | 1635 | 79 | ||
| NM_134086.4 | NP_598847.2 | 13 - 14 | 2121 | 79 | ||
| GenBank mRNA | AK034130.1 | - | 13 - 14 | 2123 | 79 | |
| AK035098.1 | - | 12 - 13 | 1140 | 79 | ||
| AK047459.1 | - | 13 - 14 | 1652 | 79 | ||
| AK049173.1 | - | 13 - 14 | 1337 | 80 | ||
| AK049294.1 | - | 13 - 14 | 1401 | 79 | ||
| AK081724.1 | - | 13 - 14 | 1637 | 79 | ||
| AK082683.1 | - | 13 - 14 | 1339 | 79 | ||
| AK162612.1 | - | 13 - 14 | 1486 | 79 | ||
| AK163914.1 | - | 8 - 9 | 739 | 79 | ||
| AK165188.1 | - | 13 - 14 | 1688 | 79 | ||
| AK204713.1 | - | 1 - 2 | 134 | 79 | ||
| BC030378.1 | - | 13 - 14 | 1337 | 5375319 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc38a1
Genomic Map