Assay Details
| Gene Symbol: Dhh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007857.4 | NP_031883.1 | 1 - 2 | 567 | 93 | |
| GenBank mRNA | AK033392.1 | - | 1 - 2 | 568 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dhh
Genomic Map