Assay Details
| Gene Symbol: Sucla2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011506.3 | NP_035636.1 | 4 - 5 | 553 | 80 | |
| GenBank mRNA | AF058955.1 | - | 3 - 4 | 420 | 80 | |
| AK005273.1 | - | 4 - 5 | 535 | 80 | ||
| AK081965.1 | - | 4 - 5 | 574 | 80 | ||
| AK088801.1 | - | 4 - 5 | 554 | 80 | ||
| AK132762.1 | - | 4 - 5 | 543 | 80 | ||
| AK150594.1 | - | 4 - 5 | 553 | 80 | ||
| AK152783.1 | - | 4 - 5 | 553 | 80 | ||
| AK153198.1 | - | 4 - 5 | 553 | 80 | ||
| AK160130.1 | - | 4 - 5 | 554 | 80 | ||
| AK160652.1 | - | 4 - 5 | 542 | 80 | ||
| BC056353.1 | - | 4 - 5 | 544 | 6808360 | 80 | |
| BC057605.1 | - | 4 - 5 | 544 | 6808360 | 80 | |
More Information
Set Membership: |
|

Sucla2
Genomic Map