Assay Details
| Gene Symbol: Sucla2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011506.3 | NP_035636.1 | 5 - 6 | 686 | 106 | |
| GenBank mRNA | AF058955.1 | - | 4 - 5 | 553 | 106 | |
| AK081965.1 | - | 5 - 6 | 707 | 106 | ||
| AK088801.1 | - | 5 - 6 | 687 | 106 | ||
| AK132762.1 | - | 5 - 6 | 676 | 106 | ||
| AK150594.1 | - | 5 - 6 | 686 | 106 | ||
| AK152783.1 | - | 5 - 6 | 686 | 106 | ||
| AK153198.1 | - | 5 - 6 | 686 | 106 | ||
| AK160130.1 | - | 5 - 6 | 687 | 106 | ||
| AK160652.1 | - | 5 - 6 | 675 | 106 | ||
| BC056353.1 | - | 5 - 6 | 677 | 6808360 | 106 | |
| BC057605.1 | - | 5 - 6 | 677 | 6808360 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sucla2
Genomic Map