Assay Details
| Gene Symbol: Sucla2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011506.3 | NP_035636.1 | 9 - 10 | 1249 | 143 | |
| GenBank mRNA | AF058955.1 | - | 8 - 9 | 1116 | 143 | |
| AK003867.1 | - | 1 - 2 | 134 | 143 | ||
| AK005273.1 | - | 9 - 10 | 1232 | 143 | ||
| AK081965.1 | - | 9 - 10 | 1270 | 143 | ||
| AK088801.1 | - | 9 - 10 | 1250 | 143 | ||
| AK132762.1 | - | 9 - 10 | 1239 | 143 | ||
| AK150594.1 | - | 9 - 10 | 1249 | 143 | ||
| AK152783.1 | - | 9 - 10 | 1249 | 143 | ||
| AK153198.1 | - | 9 - 10 | 1249 | 143 | ||
| AK160130.1 | - | 9 - 10 | 1250 | 143 | ||
| AK160652.1 | - | 9 - 10 | 1238 | 143 | ||
| BC038497.1 | - | 3 - 4 | 417 | 143 | ||
| BC052774.1 | - | 6 - 7 | 781 | 143 | ||
| BC056353.1 | - | 9 - 10 | 1240 | 6808360 | 143 | |
| BC057605.1 | - | 9 - 10 | 1240 | 6808360 | 143 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sucla2
Genomic Map