| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400008201.mm.1 |
NM_001170694.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400008201.mm.1 |
NM_001170694.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400008201.mm.1 |
NM_134083.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400008201.mm.1 |
NM_134083.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400008201.mm.1 |
TC1400001031.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400008237.mm.1 |
NM_001170694.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400008237.mm.1 |
NM_001170694.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400008237.mm.1 |
NM_134083.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400008237.mm.1 |
NM_134083.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400008237.mm.1 |
NR_033185.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400008237.mm.1 |
NR_033185.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400008237.mm.1 |
TC1400001031.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400008204.mm.1 |
NM_001170694.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400008204.mm.1 |
NM_001170694.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400008204.mm.1 |
NM_134083.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400008204.mm.1 |
NM_134083.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400008204.mm.1 |
TC1400001031.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400008231.mm.1 |
NM_001170694.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400008231.mm.1 |
NM_001170694.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400008231.mm.1 |
NM_134083.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400008231.mm.1 |
NM_134083.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400008231.mm.1 |
NR_033185.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400008231.mm.1 |
NR_033185.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400008231.mm.1 |
TC1400001031.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400008239.mm.1 |
NM_001170694.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400008239.mm.1 |
NM_001170694.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400008239.mm.1 |
NM_134083.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400008239.mm.1 |
NM_134083.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400008239.mm.1 |
NR_033185.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400008239.mm.1 |
NR_033185.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400008239.mm.1 |
TC1400001031.mm.1 |
MTA TC |