Assay Details
| Gene Symbol: Slc25a30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026232.3 | NP_080508.1 | 1 - 2 | 120 | 71 | |
| GenBank mRNA | AK015679.1 | - | 1 - 2 | 121 | 71 | |
| AK017035.1 | - | 1 - 2 | 121 | 71 | ||
| AK030950.1 | - | 1 - 2 | 89 | 71 | ||
| AK090086.1 | - | 1 - 2 | 58 | 71 | ||
| AK151334.1 | - | 1 - 2 | 89 | 71 | ||
| AK151420.1 | - | 1 - 2 | 57 | 71 | ||
| AK159122.1 | - | 1 - 2 | 118 | 71 | ||
| AK159732.1 | - | 1 - 2 | 118 | 71 | ||
| BC022676.1 | - | 1 - 2 | 60 | 4238839 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK015679.1, AK017035.1, AK151334.1, AK151420.1, AK159122.1, AK159732.1, BC022676.1, NM_026232.3 transcripts.
Set Membership: |
|

Slc25a30
Genomic Map