Assay Details
| Gene Symbol: Slc25a30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026232.3 | NP_080508.1 | 2 - 3 | 250 | 71 | |
| GenBank mRNA | AK015679.1 | - | 2 - 3 | 251 | 71 | |
| AK017035.1 | - | 2 - 3 | 251 | 71 | ||
| AK030950.1 | - | 2 - 3 | 219 | 71 | ||
| AK090086.1 | - | 2 - 3 | 188 | 71 | ||
| AK151334.1 | - | 2 - 3 | 219 | 71 | ||
| AK159122.1 | - | 2 - 3 | 248 | 71 | ||
| AK159732.1 | - | 2 - 3 | 248 | 71 | ||
| BC022676.1 | - | 2 - 3 | 190 | 4238839 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc25a30
Genomic Map