Assay Details
| Gene Symbol: Lrrc63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027581.1 | NP_081857.1 | 1 - 2 | 115 | 68 | |
| GenBank mRNA | AI592811.1 | - | 1 - 2 | 115 | 68 | |
| BC145796.1 | - | 1 - 2 | 113 | 40131099 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC145796.1, NM_027581.1 transcripts.
Set Membership: |
|

Lrrc63
Genomic Map