Assay Details
| Gene Symbol: Lrrc63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027581.1 | NP_081857.1 | 2 - 3 | 285 | 99 | |
| GenBank mRNA | AI592811.1 | - | 2 - 3 | 285 | 99 | |
| BC145796.1 | - | 2 - 3 | 283 | 40131099 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc63
Genomic Map