Assay Details
| Gene Symbol: Lrrc63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027581.1 | NP_081857.1 | 4 - 5 | 1123 | 71 | |
| GenBank mRNA | AK014850.1 | - | 3 - 4 | 855 | 71 | |
| BC145796.1 | - | 4 - 5 | 1118 | 40131099 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc63
Genomic Map