Assay Details
| Gene Symbol: Cd46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010778.3 | NP_034908.1 | 3 - 4 | 449 | 84 | |
| GenBank mRNA | AB001566.1 | - | 3 - 4 | 426 | 84 | |
| AB010919.1 | - | 3 - 4 | 446 | 84 | ||
| AB022600.1 | - | 3 - 4 | 426 | 84 | ||
| AK006632.1 | - | 3 - 4 | 450 | 84 | ||
| BC150660.1 | - | 3 - 4 | 450 | 9087571 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cd46
Genomic Map