| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400004127.mm.1 |
NM_001037221.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004127.mm.1 |
NM_001037221.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004127.mm.1 |
NM_001163433.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004127.mm.1 |
NM_001163433.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004127.mm.1 |
NM_028966.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004127.mm.1 |
NM_028966.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004127.mm.1 |
TC1400000507.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400004129.mm.1 |
NM_001037221.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004129.mm.1 |
NM_001037221.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004129.mm.1 |
NM_001163433.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004129.mm.1 |
NM_001163433.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004129.mm.1 |
NM_028966.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004129.mm.1 |
NM_028966.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004129.mm.1 |
TC1400000507.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400004125.mm.1 |
NM_001037221.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004125.mm.1 |
NM_001037221.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004125.mm.1 |
NM_001163433.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004125.mm.1 |
NM_001163433.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004125.mm.1 |
NM_028966.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004125.mm.1 |
NM_028966.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004125.mm.1 |
TC1400000507.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400004128.mm.1 |
NM_001037221.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004128.mm.1 |
NM_001037221.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004128.mm.1 |
NM_001163433.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004128.mm.1 |
NM_001163433.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004128.mm.1 |
NM_028966.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004128.mm.1 |
NM_028966.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004128.mm.1 |
TC1400000507.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400004126.mm.1 |
NM_001037221.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004126.mm.1 |
NM_001037221.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004126.mm.1 |
NM_001163433.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004126.mm.1 |
NM_001163433.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004126.mm.1 |
NM_028966.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004126.mm.1 |
NM_028966.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004126.mm.1 |
TC1400000507.mm.1 |
MTA TC |