Assay Details
| Gene Symbol: Folh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159706.1 | NP_001153178.1 | 3 - 4 | 516 | 106 | |
| NM_016770.3 | NP_058050.3 | 3 - 4 | 516 | 106 | ||
| GenBank mRNA | AF026380.1 | - | 3 - 4 | 498 | 106 | |
| AK090289.1 | - | 517 | 106 | |||
| BC119604.1 | - | 3 - 4 | 460 | 40060385 | 106 | |
| BC119605.1 | - | 3 - 4 | 460 | 40060386 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Folh1
Genomic Map