Assay Details
| Gene Symbol: Taf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277958.1 | NP_001264887.1 | 10 - 11 | 1435 | 134 | |
| NM_001277959.1 | NP_001264888.1 | 10 - 11 | 1414 | 134 | ||
| NM_021466.3 | NP_067441.1 | 10 - 11 | 1446 | 134 | ||
| GenBank mRNA | AK132811.1 | - | 10 - 11 | 1400 | 134 | |
| AK133426.1 | - | 10 - 11 | 1436 | 134 | ||
| BC061106.1 | - | 10 - 11 | 1331 | 6595990 | 134 | |
| Y09972.1 | - | 10 - 11 | 1348 | 134 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P110687 | Y09972.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P110687 | BC061106.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P110687 | BC138425.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P110687 | NM_021466.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P110687 | AK132811.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P110687 | Y09972.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P110687 | BC061106.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P110687 | BC138425.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P110687 | NM_021466.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P110687 | AK132811.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P516057 | Y09972.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P516057 | Y09972.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P516057 | Y09972.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P516057 | Y09972.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P516057 | Y09972.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P516057 | Y09972.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P516057 | Y09972.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P516057 | Y09972.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P516057 | Y09972.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P516057 | Y09972.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09574641 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09574518 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09574633 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09574620 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09574627 | - | - |
| Whole Mouse Genome G4122A | A_51_P516057 | Y09972.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P516057 | Y09972.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P516057 | Y09972.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P516057 | Y09972.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P516057 | Y09972.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P516057 | Y09972.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P516057 | Y09972.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P516057 | Y09972.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P516057 | Y09972.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P516057 | Y09972.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P618511 | Y09972.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P618511 | BC061106.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P618511 | BC138425.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P618511 | AK133426.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P618511 | NM_021466.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P1140007 | AK040747.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110687 | Y09972.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110687 | BC061106.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110687 | BC138425.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110687 | NM_021466.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110687 | AK132811.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09574627 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09574641 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09574518 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10598110 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10598108 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10598109 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419702_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10598107 | - | - |
| Mouse Genome 430 2.0 Array | 1419702_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1419702_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100014692.mm.1 | NM_001277957.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100014692.mm.1 | NM_001277958.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100014692.mm.1 | NM_001277959.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100014692.mm.1 | NM_021466.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100014692.mm.1 | TC0100001756.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100014693.mm.1 | NM_001277957.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100014693.mm.1 | NM_001277958.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100014693.mm.1 | NM_001277959.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100014693.mm.1 | NM_021466.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100014693.mm.1 | TC0100001756.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100014690.mm.1 | NM_001277958.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100014690.mm.1 | NM_001277959.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100014690.mm.1 | NM_021466.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100014690.mm.1 | TC0100001756.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100014691.mm.1 | NM_001277957.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100014691.mm.1 | NM_001277958.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100014691.mm.1 | NM_001277959.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100014691.mm.1 | NM_021466.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100014691.mm.1 | TC0100001756.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1419702_at | - | - |
| Mouse Expression Set 430 | 1419702_PM_at | - | - |
| Murine Genome U74v2 Set | 92339_PM_at | - | - |
| Murine Genome U74v2 Set | 92339_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419702_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419702_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419702_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419702_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 298174 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 106380593 | - | - |
| MouseWG-6 v2 Expression BeadChip | 2510707 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106380593 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2510707 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002510707 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1259398 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1259398 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2568415 | - | - |
| mouse-6 v1.1 (Illumina) | scl0021339.1_180-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106380593 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2510707 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0021339.1_180-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106380593 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2510707 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2640188 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2568415 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0021339.1_180-S | - | - |

Taf1a
Genomic Map