Assay Details
| Gene Symbol: Taf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277957.1 | NP_001264886.1 | 5 - 6 | 810 | 65 | |
| NM_001277958.1 | NP_001264887.1 | 5 - 6 | 799 | 65 | ||
| NM_001277959.1 | NP_001264888.1 | 5 - 6 | 778 | 65 | ||
| NM_021466.3 | NP_067441.1 | 5 - 6 | 810 | 65 | ||
| GenBank mRNA | AK132811.1 | - | 5 - 6 | 764 | 65 | |
| AK133426.1 | - | 5 - 6 | 800 | 65 | ||
| BC061106.1 | - | 5 - 6 | 695 | 6595990 | 65 | |
| BC138425.1 | - | 5 - 6 | 676 | 8861445 | 65 | |
| Y09972.1 | - | 5 - 6 | 712 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Taf1a
Genomic Map