Assay Details
| Gene Symbol: Arhgap36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081123.1 | NP_001074592.1 | 5 - 6 | 872 | 106 | |
| GenBank mRNA | AB073967.1 | - | 5 - 6 | 739 | 106 | |
| AK048167.1 | - | 5 - 6 | 761 | 106 | ||
| BC090672.1 | - | 5 - 6 | 843 | 30941825 | 106 | |
| BC145645.1 | - | 5 - 6 | 838 | 40130948 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap36
Genomic Map