Assay Details
| Gene Symbol: Cnst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146105.3 | NP_666217.2 | 5 - 6 | 817 | 63 | |
| GenBank mRNA | AK036332.1 | - | 5 - 6 | 849 | 63 | |
| AK146681.1 | - | 5 - 6 | 907 | 63 | ||
| BC039206.1 | - | 3 - 4 | 180 | 63 | ||
| BC054802.1 | - | 5 - 6 | 817 | 6837803 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cnst
Genomic Map