Assay Details
| Gene Symbol: Cnst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146105.3 | NP_666217.2 | 6 - 7 | 933 | 77 | |
| GenBank mRNA | AK036332.1 | - | 6 - 7 | 965 | 77 | |
| AK146681.1 | - | 6 - 7 | 1023 | 77 | ||
| BC039206.1 | - | 4 - 5 | 296 | 77 | ||
| BC054802.1 | - | 6 - 7 | 933 | 6837803 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cnst
Genomic Map