Assay Details
| Gene Symbol: Acox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161667.1 | NP_001155139.1 | 10 - 11 | 1492 | 61 | |
| NM_053115.2 | NP_444345.2 | 10 - 11 | 1617 | 61 | ||
| GenBank mRNA | AJ238492.1 | - | 10 - 11 | 1534 | 61 | |
| AK144243.1 | - | 10 - 11 | 1486 | 61 | ||
| BC021339.1 | - | 10 - 11 | 1496 | 5052291 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acox2
Genomic Map