Assay Details
| Gene Symbol: Acox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161667.1 | NP_001155139.1 | 11 - 12 | 1672 | 68 | |
| NM_053115.2 | NP_444345.2 | 11 - 12 | 1797 | 68 | ||
| GenBank mRNA | AJ238492.1 | - | 11 - 12 | 1714 | 68 | |
| AK144243.1 | - | 11 - 12 | 1666 | 68 | ||
| BC021339.1 | - | 11 - 12 | 1676 | 5052291 | 68 | |
More Information
Set Membership: |
|

Acox2
Genomic Map