Assay Details
| Gene Symbol: Dnase1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007870.3 | NP_031896.3 | 2 - 3 | 443 | 120 | |
| GenBank mRNA | AF047355.1 | - | 2 - 3 | 418 | 120 | |
| AK156109.1 | - | 276 | 120 | |||
| AK171843.1 | - | 2 - 3 | 276 | 120 | ||
| AK172254.1 | - | 2 - 3 | 836 | 120 | ||
| BC012671.1 | - | 2 - 3 | 396 | 4160709 | 120 | |
| U76110.1 | - | 2 - 3 | 252 | 120 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dnase1l3
Genomic Map