Assay Details
| Gene Symbol: Dnase1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007870.3 | NP_031896.3 | 3 - 4 | 529 | 99 | |
| GenBank mRNA | AF047355.1 | - | 3 - 4 | 504 | 99 | |
| AK156109.1 | - | 362 | 99 | |||
| AK171843.1 | - | 3 - 4 | 362 | 99 | ||
| AK172254.1 | - | 3 - 4 | 922 | 99 | ||
| BC012671.1 | - | 3 - 4 | 482 | 4160709 | 99 | |
| U76110.1 | - | 3 - 4 | 338 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dnase1l3
Genomic Map