Assay Details
| Gene Symbol: Sass6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289568.1 | NP_001276497.1 | 12 - 13 | 1563 | 167 | |
| NM_001289571.1 | NP_001276500.1 | 10 - 11 | 1419 | 167 | ||
| NM_028349.3 | NP_082625.2 | 11 - 12 | 1480 | 167 | ||
| GenBank mRNA | AK013338.1 | - | 12 - 13 | 1455 | 167 | |
| AK165224.1 | - | 12 - 13 | 1509 | 167 | ||
| BC050110.1 | - | 11 - 12 | 1430 | 30024366 | 167 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sass6
Genomic Map