Assay Details
| Gene Symbol: Sass6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289568.1 | NP_001276497.1 | 14 - 15 | 1776 | 79 | |
| NM_001289571.1 | NP_001276500.1 | 12 - 13 | 1632 | 79 | ||
| NM_028349.3 | NP_082625.2 | 13 - 14 | 1693 | 79 | ||
| GenBank mRNA | AK013338.1 | - | 14 - 15 | 1668 | 79 | |
| AK145076.1 | - | 12 - 13 | 1581 | 79 | ||
| AK165224.1 | - | 14 - 15 | 1722 | 79 | ||
| BC050110.1 | - | 13 - 14 | 1643 | 30024366 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sass6
Genomic Map