Assay Details
| Gene Symbol: Sass6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289568.1 | NP_001276497.1 | 15 - 16 | 1904 | 87 | |
| NM_001289571.1 | NP_001276500.1 | 13 - 14 | 1760 | 87 | ||
| NM_028349.3 | NP_082625.2 | 14 - 15 | 1821 | 87 | ||
| GenBank mRNA | AK145076.1 | - | 13 - 14 | 1709 | 87 | |
| AK165224.1 | - | 15 - 16 | 1850 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sass6
Genomic Map