Assay Details
| Gene Symbol: Sass6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289568.1 | NP_001276497.1 | 4 - 5 | 448 | 152 | |
| NM_001289571.1 | NP_001276500.1 | 2 - 3 | 304 | 152 | ||
| NM_028349.3 | NP_082625.2 | 3 - 4 | 365 | 152 | ||
| GenBank mRNA | AK013338.1 | - | 4 - 5 | 340 | 152 | |
| AK145076.1 | - | 2 - 3 | 253 | 152 | ||
| AK165224.1 | - | 4 - 5 | 394 | 152 | ||
| BB617231.1 | - | 3 - 4 | 367 | 152 | ||
| BC050110.1 | - | 3 - 4 | 315 | 30024366 | 152 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001289568.1 transcripts.
Set Membership: |
|

Sass6
Genomic Map