Assay Details
| Gene Symbol: Sass6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289568.1 | NP_001276497.1 | 7 - 8 | 790 | 67 | |
| NM_001289571.1 | NP_001276500.1 | 5 - 6 | 646 | 67 | ||
| NM_028349.3 | NP_082625.2 | 6 - 7 | 707 | 67 | ||
| GenBank mRNA | AK013338.1 | - | 7 - 8 | 682 | 67 | |
| AK145076.1 | - | 5 - 6 | 595 | 67 | ||
| AK165224.1 | - | 7 - 8 | 736 | 67 | ||
| BC050110.1 | - | 6 - 7 | 657 | 30024366 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sass6
Genomic Map