Assay Details
| Gene Symbol: Sass6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289568.1 | NP_001276497.1 | 8 - 9 | 908 | 68 | |
| NM_001289571.1 | NP_001276500.1 | 6 - 7 | 764 | 68 | ||
| NM_028349.3 | NP_082625.2 | 7 - 8 | 825 | 68 | ||
| GenBank mRNA | AK013338.1 | - | 8 - 9 | 800 | 68 | |
| AK165224.1 | - | 8 - 9 | 854 | 68 | ||
| BC050110.1 | - | 7 - 8 | 775 | 30024366 | 68 | |
More Information
Set Membership: |
|

Sass6
Genomic Map