Assay Details
| Gene Symbol: Sass6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289568.1 | NP_001276497.1 | 10 - 11 | 1294 | 144 | |
| NM_001289571.1 | NP_001276500.1 | 8 - 9 | 1150 | 144 | ||
| NM_028349.3 | NP_082625.2 | 9 - 10 | 1211 | 144 | ||
| GenBank mRNA | AK013338.1 | - | 10 - 11 | 1186 | 144 | |
| AK145076.1 | - | 8 - 9 | 1099 | 144 | ||
| AK165224.1 | - | 10 - 11 | 1240 | 144 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sass6
Genomic Map