Assay Details
| Gene Symbol: Syt14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301370.1 | NP_001288299.1 | 5 - 6 | 853 | 112 | |
| NM_181546.3 | NP_853524.1 | 5 - 6 | 853 | 112 | ||
| GenBank mRNA | AB102947.1 | - | 5 - 6 | 776 | 112 | |
| BC141262.1 | - | 5 - 6 | 795 | 9055874 | 112 | |
| BC145602.1 | - | 5 - 6 | 795 | 9054168 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Syt14
Genomic Map