| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300025072.mm.1 |
NM_001093752.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300025072.mm.1 |
NM_001093752.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300025072.mm.1 |
NM_001093753.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300025072.mm.1 |
NM_001093753.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300025072.mm.1 |
TC0300003196.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300025073.mm.1 |
NM_001093752.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300025073.mm.1 |
NM_001093752.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300025073.mm.1 |
NM_001093753.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300025073.mm.1 |
NM_001093753.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300025073.mm.1 |
TC0300003196.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300025076.mm.1 |
TC0300003196.mm.1 |
MTA TC |