Assay Details
| Gene Symbol: Lrrc40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024194.6 | NP_077156.2 | 2 - 3 | 384 | 163 | |
| GenBank mRNA | AK011726.1 | - | 2 - 3 | 365 | 163 | |
| AK017814.1 | - | 2 - 3 | 353 | 163 | ||
| AK034978.1 | - | 364 | 163 | |||
| AK148748.1 | - | 2 - 3 | 343 | 163 | ||
| AK163288.1 | - | 2 - 3 | 352 | 163 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc40
Genomic Map