Assay Details
| Gene Symbol: Lrrc40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289524.1 | NP_001276453.1 | 4 - 5 | 581 | 72 | |
| NM_001289525.1 | NP_001276454.1 | 2 - 3 | 328 | 72 | ||
| NM_024194.6 | NP_077156.2 | 4 - 5 | 584 | 72 | ||
| GenBank mRNA | AK011726.1 | - | 4 - 5 | 565 | 72 | |
| AK017814.1 | - | 4 - 5 | 553 | 72 | ||
| AK034978.1 | - | 564 | 72 | |||
| AK148748.1 | - | 4 - 5 | 543 | 72 | ||
| AK152244.1 | - | 2 - 3 | 309 | 72 | ||
| AK163288.1 | - | 4 - 5 | 552 | 72 | ||
| AK170052.1 | - | 4 - 5 | 561 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK152244.1, AK170052.1, NM_001289524.1, NM_001289525.1 transcripts.
Set Membership: |
|

Lrrc40
Genomic Map