Assay Details
| Gene Symbol: Lrrc40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289524.1 | NP_001276453.1 | 7 - 8 | 1019 | 154 | |
| NM_001289525.1 | NP_001276454.1 | 5 - 6 | 766 | 154 | ||
| NM_024194.6 | NP_077156.2 | 7 - 8 | 1022 | 154 | ||
| GenBank mRNA | AK011726.1 | - | 7 - 8 | 1003 | 154 | |
| AK017814.1 | - | 7 - 8 | 991 | 154 | ||
| AK034978.1 | - | 1002 | 154 | |||
| AK148748.1 | - | 7 - 8 | 982 | 154 | ||
| AK152244.1 | - | 5 - 6 | 747 | 154 | ||
| AK163288.1 | - | 7 - 8 | 990 | 154 | ||
| AK170052.1 | - | 7 - 8 | 999 | 154 | ||
| BC002279.1 | - | 3 - 4 | 411 | 3497400 | 154 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc40
Genomic Map