Assay Details
| Gene Symbol: Lrrc40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289524.1 | NP_001276453.1 | 8 - 9 | 1105 | 121 | |
| NM_001289525.1 | NP_001276454.1 | 6 - 7 | 852 | 121 | ||
| NM_024194.6 | NP_077156.2 | 8 - 9 | 1108 | 121 | ||
| GenBank mRNA | AK011726.1 | - | 8 - 9 | 1089 | 121 | |
| AK017814.1 | - | 8 - 9 | 1077 | 121 | ||
| AK034978.1 | - | 1088 | 121 | |||
| AK148748.1 | - | 8 - 9 | 1068 | 121 | ||
| AK152244.1 | - | 6 - 7 | 833 | 121 | ||
| AK163288.1 | - | 8 - 9 | 1076 | 121 | ||
| AK170052.1 | - | 8 - 9 | 1085 | 121 | ||
| BC002279.1 | - | 4 - 5 | 497 | 3497400 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc40
Genomic Map