Assay Details
| Gene Symbol: Mctp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024703.1 | NP_001019874.1 | 10 - 11 | 1491 | 93 | |
| GenBank mRNA | BC086658.1 | - | 10 - 11 | 1491 | 30546156 | 93 |
More Information
Updated Mapping Information
Set Membership: |
|

Mctp2
Genomic Map