Assay Details
| Gene Symbol: Chrna7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007390.3 | NP_031416.3 | 3 - 4 | 292 | 90 | |
| GenBank mRNA | AF325348.1 | - | 2 - 3 | 89 | 90 | |
| AK034208.1 | - | 3 - 4 | 336 | 90 | ||
| AK157809.1 | - | 3 - 4 | 289 | 90 | ||
| AY574266.1 | - | 3 - 4 | 242 | 90 | ||
| AY851289.1 | - | 117 | 90 | |||
| L37663.1 | - | 3 - 4 | 292 | 90 | ||
More Information
Set Membership: |
|

Chrna7
Genomic Map