Assay Details
| Gene Symbol: Trpm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039104.2 | NP_001034193.2 | 10 - 11 | 1527 | 95 | |
| NM_018752.3 | NP_061222.3 | 10 - 11 | 1420 | 95 | ||
| GenBank mRNA | AK149290.1 | - | 10 - 11 | 1449 | 95 | |
| AK164395.1 | - | 10 - 11 | 1330 | 95 | ||
| AY180104.2 | - | 10 - 11 | 1527 | 95 | ||
| BC082560.1 | - | 10 - 11 | 1296 | 95 | ||
| BC085168.1 | - | 9 - 10 | 1099 | 95 | ||
More Information
Set Membership: |
|

Trpm1
Genomic Map