Assay Details
| Gene Symbol: Lpar3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022983.4 | NP_075359.1 | 1 - 2 | 191 | 158 | |
| GenBank mRNA | AK042084.1 | - | 1 - 2 | 168 | 158 | |
| AK042255.1 | - | 1 - 2 | 179 | 158 | ||
| AK132289.1 | - | 1 - 2 | 133 | 158 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK042084.1, NM_022983.4 transcripts.
Set Membership: |
|

Lpar3
Genomic Map