Assay Details
| Gene Symbol: Bmpr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277216.1 | NP_001264145.1 | 5 - 6 | 516 | 65 | |
| NM_001277217.1 | NP_001264146.1 | 5 - 6 | 542 | 65 | ||
| NM_001277218.1 | NP_001264147.1 | 4 - 5 | 555 | 65 | ||
| NM_001277220.1 | NP_001264149.1 | 5 - 6 | 690 | 65 | ||
| NM_007560.4 | NP_031586.1 | 4 - 5 | 709 | 65 | ||
| GenBank mRNA | AK086130.1 | - | 5 - 6 | 502 | 65 | |
| AK140158.1 | - | 5 - 6 | 614 | 65 | ||
| AK162844.1 | - | 4 - 5 | 390 | 65 | ||
| BC065106.1 | - | 4 - 5 | 490 | 30357130 | 65 | |
| BC065143.1 | - | 4 - 5 | 696 | 30629345 | 65 | |
| BC138638.1 | - | 4 - 5 | 436 | 8861658 | 65 | |
| BC138640.1 | - | 4 - 5 | 436 | 8861660 | 65 | |
| CV557650.1 | - | 4 - 5 | 553 | 65 | ||
| Z23143.1 | - | 5 - 6 | 534 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bmpr1b
Genomic Map