Assay Details
| Gene Symbol: Bmpr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277216.1 | NP_001264145.1 | 10 - 11 | 1421 | 68 | |
| NM_001277217.1 | NP_001264146.1 | 10 - 11 | 1447 | 68 | ||
| NM_001277218.1 | NP_001264147.1 | 9 - 10 | 1460 | 68 | ||
| NM_001277220.1 | NP_001264149.1 | 10 - 11 | 1595 | 68 | ||
| NM_007560.4 | NP_031586.1 | 9 - 10 | 1614 | 68 | ||
| GenBank mRNA | AK086130.1 | - | 10 - 11 | 1407 | 68 | |
| AK140158.1 | - | 10 - 11 | 1519 | 68 | ||
| AK162844.1 | - | 9 - 10 | 1295 | 68 | ||
| BC065106.1 | - | 9 - 10 | 1395 | 30357130 | 68 | |
| BC065143.1 | - | 9 - 10 | 1601 | 30629345 | 68 | |
| BC138638.1 | - | 9 - 10 | 1341 | 8861658 | 68 | |
| BC138640.1 | - | 9 - 10 | 1341 | 8861660 | 68 | |
| Z23143.1 | - | 10 - 11 | 1439 | 68 | ||
More Information
Set Membership: |
|

Bmpr1b
Genomic Map