Assay Details
| Gene Symbol: Pkn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178654.4 | NP_848769.2 | 18 - 19 | 2763 | 94 | |
| GenBank mRNA | AK049713.1 | - | 18 - 19 | 2750 | 94 | |
| AK054281.1 | - | 4 - 5 | 367 | 94 | ||
| AK083425.1 | - | 13 - 14 | 1561 | 94 | ||
| AK133298.1 | - | 18 - 19 | 2752 | 94 | ||
| AK171092.1 | - | 18 - 19 | 2694 | 94 | ||
| BC052073.1 | - | 19 - 20 | 2686 | 6400440 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pkn2
Genomic Map