Assay Details
| Gene Symbol: Fah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010176.4 | NP_034306.2 | 12 - 13 | 1280 | 59 | |
| GenBank mRNA | AI848212.1 | - | 2 - 3 | 59 | ||
| AK143759.1 | - | 12 - 13 | 1109 | 59 | ||
| AK158808.1 | - | 12 - 13 | 1166 | 59 | ||
| BC010767.1 | - | 12 - 13 | 1065 | 4242421 | 59 | |
| M84145.1 | - | 12 - 13 | 1106 | 59 | ||
| Z11774.1 | - | 12 - 13 | 1092 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fah
Genomic Map