Assay Details
| Gene Symbol: Fah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010176.4 | NP_034306.2 | 4 - 5 | 582 | 66 | |
| GenBank mRNA | AK143759.1 | - | 4 - 5 | 411 | 66 | |
| AK158808.1 | - | 4 - 5 | 468 | 66 | ||
| BC010767.1 | - | 4 - 5 | 367 | 4242421 | 66 | |
| M84145.1 | - | 4 - 5 | 408 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fah
Genomic Map