Assay Details
| Gene Symbol: Arnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007488.3 | NP_031514.3 | 8 - 9 | 1031 | 69 | |
| GenBank mRNA | AK032628.1 | - | 8 - 9 | 1012 | 69 | |
| BC054546.1 | - | 8 - 9 | 952 | 6409636 | 69 | |
More Information
Set Membership: |
|

Arnt2
Genomic Map